Skip to Content
Merck
All Photos(1)

Key Documents

SML0735

Sigma-Aldrich

Tropodithietic acid

≥98% (HPLC)

Synonym(s):

3-Oxo-8,9-dithiabicyclo[5.2.0]nona-1,4,6-triene-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4O3S2
CAS Number:
Molecular Weight:
212.25
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

Roseobacter gallaeciensis

Assay

≥98% (HPLC)

form

powder

color

faintly yellow to dark yellow

solubility

DMSO: 5 mg/mL, clear (warmed)

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

Tropodithietic acid is a broad spectrum tropolone antibiotic with activity against both Gram-negative and Gram-positive bacteria.
Tropodithietic acid is a broad spectrum tropolone antibiotic.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natsuko Miura et al.
Marine biotechnology (New York, N.Y.), 21(1), 1-8 (2018-09-09)
The coral microbiome has attracted increased attention because of its potential roles in host protection against deadly diseases. However, little is known about the role of coral-associated bacteria against the temperature-dependent opportunistic pathogen Vibrio coralliilyticus. In this study, we tested

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service