Skip to Content
Merck
All Photos(1)

Key Documents

SML0509

Sigma-Aldrich

JK-P3

≥98% (HPLC)

Synonym(s):

3,4-Dimethoxy-N-(5-phenyl-1H-pyrazol-3-yl)-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17N3O3
CAS Number:
Molecular Weight:
323.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

COc1ccc(cc1OC)C(=O)Nc2cc([nH]n2)-c3ccccc3

InChI

1S/C18H17N3O3/c1-23-15-9-8-13(10-16(15)24-2)18(22)19-17-11-14(20-21-17)12-6-4-3-5-7-12/h3-11H,1-2H3,(H2,19,20,21,22)

InChI key

QAZJUVDICQNITG-UHFFFAOYSA-N

Biochem/physiol Actions

JK-P3 is a VEGFR-2 inhibitor.
JK-P3 is a pyrazole-based inhibitor of VEGFR-2 (IC50 = 7.8 μM). JK-P3 inhibits FGFR 1/3 kinase activity in vitro, but has no effect on FGFR signaling in cell-based assays. The compound blocks wound healing and tube formation in HUVEC without effecting endothelial cell proliferation.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the FGFR page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to FGFR for your research needs.

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service