Skip to Content
Merck
All Photos(1)

Key Documents

SML0254

Sigma-Aldrich

4-MDM

≥98% (HPLC)

Synonym(s):

4-Methoxydiphenylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14O
CAS Number:
Molecular Weight:
198.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

oil (Low melting solid)

color

colorless

storage temp.

room temp

SMILES string

COc1ccc(Cc2ccccc2)cc1

InChI

1S/C14H14O/c1-15-14-9-7-13(8-10-14)11-12-5-3-2-4-6-12/h2-10H,11H2,1H3

InChI key

GQLYCRTUQGSDSM-UHFFFAOYSA-N

Biochem/physiol Actions

4-MDM augments the aminopeptidase activity of LTA4 hydrolase without affecting the conversion of LTA4 to LTB4.
4-MDM augments the aminopeptidase activity of LTA4 hydrolase without affecting the conversion of LTA4 to LTB4. Treatment with 4-MDM protects mouse lungs from intranasal elastase-induced pulmonary emphysema.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shin Numao et al.
Scientific reports, 7(1), 13591-13591 (2017-10-21)
Leukotriene A4 Hydrolase (LTA4H) is a bifunctional zinc metalloenzyme that comprises both epoxide hydrolase and aminopeptidase activity, exerted by two overlapping catalytic sites. The epoxide hydrolase function of the enzyme catalyzes the biosynthesis of the pro-inflammatory lipid mediator leukotriene (LT)
Caroline M Low et al.
Scientific reports, 7, 44449-44449 (2017-03-18)
The pro-inflammatory mediator leukotriene B

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service