Skip to Content
Merck
All Photos(1)

Key Documents

SMB01060

Sigma-Aldrich

Phellamurin

≥90% (LC/MS-ELSD)

Synonym(s):

8-Prenyldihydrokaempferol 7-glucoside, Fellavin, Flacoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30O11
CAS Number:
Molecular Weight:
518.51
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

518.51

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)Oc2c(c3c(c(c2)O)C(=O)[C@@H]([C@H](O3)c4ccc(cc4)O)O)CC=C(C)C

InChI

1S/C26H30O11/c1-11(2)3-8-14-16(35-26-23(34)21(32)19(30)17(10-27)36-26)9-15(29)18-20(31)22(33)24(37-25(14)18)12-4-6-13(28)7-5-12/h3-7,9,17,19,21-24,26-30,32-34H,8,10H2,1-2H3/t17-,19-,21+,22+,23-,24-,26-/m1/s1

InChI key

GRDZTDZJQRPNCN-YIANMRPHSA-N

General description

Phellamurin, a trihydroxyflavanone, is a bioactive natural compound commonly sourced from Phellodendron amurense, Phellodendron chinense, and Commiphora africana plants. Current research indicates that this plant-derived metabolite acts as an inhibitor and may possess a range of biological activities, including antioxidant, anti-tumor, anti-inflammatory, and antimicrobial properties.

Application

Phellamurin is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Phellamurin induces cells apoptosis, has anti-tumor activity, exhibits inhibition of intestinal P-glycoprotein and it has a role as a metabolite.

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service