Skip to Content
Merck
All Photos(4)

Key Documents

SAB5702707

Sigma-Aldrich

Anti-Nectin 2/CD112 Antibody, clone 3H0G2, Rabbit Monoclonal

Synonym(s):

CD112, HVEB, PRR2, PVRL2, PVRR2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
HUF 155,000.00

HUF 155,000.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
HUF 155,000.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

HUF 155,000.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

3H0G2, monoclonal

form

liquid

mol wt

72 kDa

species reactivity

human, rat, rat

concentration

0.42 mg/mL

technique(s)

immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

TPKAKLEAQEMPSQLFTLGASEHSPLKTPYFDAGASCTEQEMPRYHELPTLEERSGPLHPGATSLGSPIPVPPGPPAVEDVSLDLEDEEGEEEEEYLDKINPIYDALSYSSPSDSYQGKGFVMSRAMYV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NECTIN2(5819)

General description

This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq, Jul 2008]

Immunogen

PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 410-538 of human Nectin 2/CD112 (Q92692).

Application

WB, IHC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service