Skip to Content
Merck
All Photos(3)

Key Documents

SAB5702618

Sigma-Aldrich

Anti-ACTN2 Antibody

rabbit monoclonal, 2P7E6

Synonym(s):

CMD1AA, CMH23

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
HUF 155,000.00

HUF 155,000.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
HUF 155,000.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

HUF 155,000.00


Please contact Customer Service for Availability

Product Name

Anti-ACTN2 Antibody, clone 2P7E6, Rabbit Monoclonal,

biological source

rabbit

Quality Level

material

colorless

clone

2P7E6, monoclonal

form

liquid

mol wt

104 kDa

species reactivity

rat, human, rat

concentration

0.34 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MNQIEPGVQYNYVYDEDEYMIQEEEWDRDLLLDPAWEKQQRKTFTAWCNSHLRKAGTQIENIEEDFRNGLKLMLLLEVISGERLPKPDRGKMRFHKIANV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTN2(88)

General description

Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a muscle-specific, alpha actinin isoform that is expressed in both skeletal and cardiac muscles. Several transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, May 2013]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ACTN2 (P35609).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service