Skip to Content
Merck
All Photos(2)

Key Documents

SAB5702206

Sigma-Aldrich

Anti-PHF8 Antibody, clone 2H9D4, Rabbit Monoclonal

Synonym(s):

JHDM1F, KDM7B, MRXSSD, ZNF422

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
HUF 155,000.00

HUF 155,000.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
HUF 155,000.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

HUF 155,000.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

2H9D4, monoclonal

form

liquid

mol wt

140 kDa

species reactivity

human, rat, rat

concentration

1.00 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

TVSNSPASQRTPGKRPIKRPAYWRTESEEEEENASLDEQDSLGACFKDAEYIYPSLESDDDDPALKSRPKKKKNSDDAPWSPKARVTPTLPKQDRPVREGT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PHF8(23133)

General description

The protein encoded by this gene is a histone lysine demethylase that preferentially acts on histones in the monomethyl or dimethyl states. The encoded protein requires Fe(2+) ion, 2-oxoglutarate, and oxygen for its catalytic activity. Defects in this gene are a cause of mental retardation syndromic X-linked Siderius type (MRXSSD). Four transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, May 2010]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 800-900 of human PHF8 (Q9UPP1).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service