Skip to Content
Merck
All Photos(2)

Key Documents

SAB5702116

Sigma-Aldrich

Anti-Caspase-6 Antibody, clone 2W4G2, Rabbit Monoclonal

Synonym(s):

CASP6,MCH2,caspase-6,Caspase-6 p18,Caspase-6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
HUF 155,000.00

HUF 155,000.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
HUF 155,000.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

HUF 155,000.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

2W4G2, monoclonal

form

liquid

mol wt

34 kDa

species reactivity

mouse, mouse

concentration

0.444 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

IHEVSTVSHADADCFVCVFLSHGEGNHIYAYDAKIEIQTLTGLFKGDKCHSLVGKPKIFIIQACRGNQHDVPVIPLDVVDNQTEKLDTNITEVDAASVYTL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CASP6(839)

General description

This gene encodes a member of the cysteine-aspartic acid protease (caspase) family of enzymes. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic acid residues to produce two subunits, large and small, that dimerize to form the active enzyme. This protein is processed by caspases 7, 8 and 10, and is thought to function as a downstream enzyme in the caspase activation cascade. Alternative splicing of this gene results in multiple transcript variants that encode different isoforms. [provided by RefSeq, Oct 2015]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Caspase-6 (P55212).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service