Skip to Content
Merck
All Photos(1)

Key Documents

MTP001

Sigma-Aldrich

MITOCHONDRIAL PROTEIN IP KIT

Sufficient for 50 tests

Synonym(s):

mitochondrial protein immunoprecipitation kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

form

liquid

technique(s)

immunoprecipitation (IP): suitable

storage temp.

−20°C

General description

The mitochondrial protein immunoprecipitation (IP) kit contains a ready-to-use mitochondria protein IP buffer, optimized for immunoprecipitation (IP and co-IP) using mitochondria and mitochondrial extracts. The buffer is a gentle formulation, which maintains the stability of mitochondrial complexes.

Application

MITOCHONDRIAL PROTEIN IP KIT has been used to perform immunoprecipitation[1] and co-immunoprecipitation (Co-IP) of mitochondrial proteins.[2]

Biochem/physiol Actions

The mitochondrial protein immunoprecipitation (IP) kit also provides choices of detergents, N-dodecyl-β-D-maltoside, Triton X-100, and digitonin, to achieve different stringency conditions for protein-protein interaction studies. Triton X-100 is the most commonly used detergent, especially for membrane protein solubilization. The mitochondrial protein immunoprecipitation (IP) kit may be used for the following:
  • optimized for compatibility with immunoprecipitation (IP and co-IP) and pull-down using tagged proteins
  • gentle formulation for maintenance of stable mitochondrial complexes
  • compatible with sodium dodecyl sulfate–polyacrylamide gel electrophoresis (SDS PAGE), 2D gel, native gel, and mass spectrometry
  • functional assays and enzymatic assays

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinchul Kim et al.
Cancer cell, 35(2), 191-203 (2019-02-05)
The tumor suppressor p53 is somatically mutated in half of all human cancers. Paradoxically, the wild-type p53 (WTp53) is often retained in certain human cancers, such as hepatocarcinoma (HCC). We discovered a physiological and oncogenic role of WTp53 in suppressing
Rongli Zhang et al.
Journal of molecular and cellular cardiology, 112, 64-73 (2017-09-09)
Heart failure is associated with mitochondrial dysfunction so that restoring or improving mitochondrial health is of therapeutic importance. Recently, reduction in NAD+ levels and NAD+-mediated deacetylase activity has been recognized as negative regulators of mitochondrial function. Using a cardiac specific

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service