Skip to Content
Merck
All Photos(1)

Key Documents

MT30671549

Sigma-Aldrich

Mettler Toledo SevenDirect SD50 pH / Ion Meter LowVol Kit

electrode, InLab® Micro Pro-ISM

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
HUF 812,000.00

HUF 812,000.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
HUF 812,000.00

About This Item

UNSPSC Code:
41115603
NACRES:
NB.61

HUF 812,000.00


Please contact Customer Service for Availability

AC/DC input

, universal plug set
220 V / 240 V

feature

electrode InLab® Micro Pro-ISM

packaging

pack of 1 kit

manufacturer/tradename

Mettler-Toledo 30671549

concentration measuring range

-2.000-20.000 pX, resolution: 3 or 4 digits (ion)
0-999,999 mg/L (ion)

mV measuring range

-2,000.000-2,000.000 mV, resolution: 1; 0.1

pH measuring range

accuracy: ±0.002, resolution: 0.1
resolution: 0.001
resolution: 0.01

temperature measuring range

-30-130 °C, resolution: 0.1 °C

General description

Mastered in no Time
Large color touchscreen provides direct measurement and easy data entry. Instructions guide users through the sensor calibration process.
Tracks your Records
The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
Tough Enough
Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
pH sensor for low sample volumes
InLab Micro-Pro ISM is a pH/temperature sensor for measurements in sample volumes down to 100 microliters. The compact design includes a temperature probe while keeping the shaft diameter at 5 mm.
Electrode Arm for Efficient Operation
The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy. Dedicated positions for cleaning and storage support correct sensor handling. Vertical and horizontal movements facilitate precise positioning, even with one hand.
Protect Critical Settings
A two-level user management protects the most important settings. Operators retain the flexibility to individually set preferences like language and screen brightness.
Display Adapts to Your Needs
View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.

Application

pH and Ion measurement

Features and Benefits

  • Large color touchscreen provides direct measurement and easy data entry.
  • The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
  • Smart design and cover keep the instrument safe from dust and spills
  • The pH/temperature sensor for measurements in sample volumes down to 100 microliters and with its 5 mm shaft diameter it fits the smallest tubes.
  • Electrode arm for efficient operation
  • Protect critical settings Protected with two-level user management
  • Display adapts to your needs to maximize readability

Components

Includes: Meter;Sensor; an efficient electrode arm; power supply; pH buffer sachets for initial calibration.

Legal Information

InLab is a registered trademark of Mettler-Toledo, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the difference between MT30671549 et MT30671561? Thanks?

    1 answer
    1. The main difference between MT30671549 and MT30671561 is the specific model of the pH/ion meter in each kit. MT30671549 is the Mettler Toledo SevenDirect SD50 pH/Ion Meter LowVol Kit. The SD50 model offers pH and ion measurement capabilities and is designed for low-volume samples.

      MT30671561 is the Mettler Toledo SevenDirect SD20 pH/ORP Meter LowVol Kit. The SD20 model provides pH and ORP (Oxidation-Reduction Potential) measurement capabilities and is also designed for low-volume samples. The key distinction between these two kits lies in the exact meter model (SD50 versus SD20) and the specific measurement functionalities they offer (ion measurement in the SD50 versus ORP measurement in the SD20).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service