Skip to Content
Merck
All Photos(1)

Key Documents

MAK513

Sigma-Aldrich

Maltose Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Maltose Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
HUF 167,000.00

HUF 167,000.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
HUF 167,000.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

HUF 167,000.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

application(s)

cosmetics
food and beverages

detection method

colorimetric
fluorometric

relevant disease(s)

endocrinological disorders, diabetes

shipped in

wet ice

storage temp.

−20°C

General description

Maltose is a disaccharide, composed of two glucose units linked by an α bond. It is produced from the hydrolysis of glycogen or starch, serving as a source of energy for plants and animals. Maltose can be found in foods such as grains, and other processed products.

Features and Benefits

  • Linear detection range 2 to 500 µM maltose for the colorimetric assay and 1 to 50 µM for the fluorometric assay.
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for maltose determination in various biological samples such as serum, urine, food and beverages, etc.

Principle

The Maltose Assay Kit provides a simple, one step assay for measuring maltose. In this assay, maltose is converted to two glucoses, which are then oxidized to form a colored product. The color intensity of the product at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 585 nm is directly proportional to maltose concentration in the sample.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service