Skip to Content
Merck
All Photos(1)

Key Documents

GW22611

Sigma-Aldrich

Anti-Interferon α2 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-IFN-alpha-2, Anti-IFN-alphaA, Anti-IFNA, Anti-IFNA2B, Anti-leIF A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-22611

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFNA2(3440)

General description

IFN α 2 is a protein that belongs to family of pleiotropic cytokines produced by macrophages in response to pathogens. It plays a crucial role in antiviral activities. Anti-interferon α2 antibody (1:200) can be used for cell or tissue staining. Chicken anti- interferon α2 antibody reacts specifically with human recombinant interferon protein α 2.

Immunogen

Immunogen Sequence: GI # 11067751, sequence 25-186
Recombinant interferon, α 2

Application

Anti-interferon α2 antibody can be used in western blotting and indirect ELISA.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J de Veer et al.
Journal of leukocyte biology, 69(6), 912-920 (2001-06-19)
Interferons (IFNs) are a family of multifunctional cytokines that activate transcription of subsets of genes. The gene products induced by IFNs are responsible for IFN antiviral, antiproliferative, and immunomodulatory properties. To obtain a more comprehensive list and a better understanding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service