Skip to Content
Merck
All Photos(1)

Key Documents

GEUS78225

illustra ExoProStar

Cytiva US78225, sufficient for 5000 reactions

Synonym(s):

Exosome Purification Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
HUF 1,397,000.00

HUF 1,397,000.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
HUF 1,397,000.00

About This Item

UNSPSC Code:
41106300
NACRES:
NA.52

HUF 1,397,000.00


Please contact Customer Service for Availability

usage

sufficient for 5000 reactions

manufacturer/tradename

Cytiva US78225

shipped in

dry ice

storage temp.

−20°C

General description

Enzymatic PCR clean up technology with illustra™ Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs. For maximum flexibility comes in two separate tubes.illustra™ ExoProStar is a new name for this PCR clean-up technology. No change in the formulation, so you get the same great product, and same great benefits! The ExoProStar PCR and Sequence Reaction Clean-Up Kit updates existing enzymatic PCR clean up technology with illustra™ Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs. illustra™ ExoProStar improves digestion efficiency with no degradation of the target PCR product. Your sample is completely intact and ready for immediate use in manual or automated processes.

Features and Benefits

  • Enzymes optimized to work together for high efficiency removal of unincorporated primers and nucleotides.
  • Enzymes provided in two separate tubes; just two simple pipetting steps are needed to prepare the reaction.
  • Fast 30 min protocol.
  • Scalable for different reaction sizes.
  • No loss of PCR product.
  • Easy to automate.
  • Complete heat inactivation of the enzymes within 15 min.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

ExoProStar is a trademark of Cytiva
illustra is a trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service