Skip to Content
Merck
All Photos(1)

Key Documents

G0148

Sigma-Aldrich

Grace′s amino acids solution- Modified for TC-100

10 ×, suitable for insect cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352209

sterility

sterile-filtered

form

solution

concentration

10 ×

technique(s)

cell culture | insect: suitable

impurities

endotoxin, tested

shipped in

ambient

storage temp.

2-8°C

General description

This is a 10X stock solution of Grace′s amino acids modified for use in TC-100. Grace′s medium was originally formulated to support the growth of cells derived from the Australian emperor gum moth, Antherea eucalypti. The medium is a modification of Wyatt′s medium to more closely resemble Antherea hemolymph. The unparalled success achieved by Grace in maintaining long term cultures of a variety of insect cells led to a resurgence in the field of insect tissue culture and wide usage of the medium. Many modifications of this medium have been developed since its introduction.

Formula variant

With L-serine. Without β-alanine, L-glutamine and D-serine.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service