EPM1196-9906
Platform for Eppendorf® New Brunswick™ Innova® 5000 Platform Shaker
for 4 L Erlenmeyer flasks, dedicated, interchangeable
Select a Size
HUF 723,000.00
Select a Size
About This Item
HUF 723,000.00
Recommended Products
feature
dedicated
for 4 L Erlenmeyer flasks
interchangeable
manufacturer/tradename
Eppendorf® M1196-9906 (NA)
size
32.5 in. × 26.75 in.
capacity
(9 x 4 L Erlenmeyer flasks)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service