Skip to Content
Merck
All Photos(1)

Key Documents

EHU134761

Sigma-Aldrich

MISSION® esiRNA

targeting human CRTAC1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
HUF 75,800.00
50 μG
HUF 135,000.00

HUF 75,800.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
HUF 75,800.00
50 μG
HUF 135,000.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

HUF 75,800.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGTAATCCCACCCAGCTCAACTATGGTGTGGCAGTTACTGATGTGGACCATGATGGGGACTTTGAGATCGTCGTGGCGGGGTACAATGGACCCAACCTGGTTCTGAAGTATGACCGGGCCCAGAAGCGGCTGGTGAACATCGCGGTCGATGAGCGCAGCTCACCCTACTACGCGCTGCGGGACCGGCAGGGGAACGCCATCGGGGTCACAGCCTGCGACATCGACGGGGACGGCCGGGAGGAGATCTACTTCCTCAACACCAATAATGCCTTCTCGGGGGTGGCCACGTACACCGACAAGTTGTTCAAGTTCCGCAATAACCGGTGGGAAGACATCCTGAGCGATGAGGTCAACGTGGCCCGTGGTGTGGCCAGCCTCTTTGCCGGACGCTCTGTGGCCTGTGTGGACAGAAAGGGCTCTGGACGCTACT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EHU116701EHU003941EHU031991
Gene Information

human ... CRTAC1(55118), CRTAC1(55118)

Gene Information

human ... EXD2(55218), EXDL2(55218)

Gene Information

human ... WDR60(55112), WDR60(55112)

Gene Information

human ... UBR7(55148), UBR7(55148)

Ensembl | human accession no.

ENSG00000095713

Ensembl | human accession no.

ENSG00000081177

Ensembl | human accession no.

ENSG00000126870

Ensembl | human accession no.

ENSG00000012963

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

NCBI accession no.

NM_018058

NCBI accession no.

NM_018199

NCBI accession no.

-

NCBI accession no.

NM_175748

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Sun et al.
DNA and cell biology, 39(4), 513-521 (2020-01-31)
The apoptosis of human lens epithelial cells (HLECs) is a characteristic change that occurs during the development of cataracts. Ultraviolet B (UVB) is known to induce the generation of reactive oxygen species (ROS) and apoptosis in HLECs, and thus cause

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service