Skip to Content
Merck
All Photos(1)

Key Documents

EHU020611

Sigma-Aldrich

MISSION® esiRNA

targeting human MUS81

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
HUF 75,800.00
50 μG
HUF 135,000.00

HUF 75,800.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
HUF 75,800.00
50 μG
HUF 135,000.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

HUF 75,800.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGTCGGTGCGAGAAGTGTTTGCCCGGCAGCTGATGCAGGTGCGCGGAGTGAGTGGGGAGAAGGCAGCAGCCCTGGTGGATCGATACAGCACCCCTGCCAGCCTCCTGGCCGCCTATGATGCCTGTGCCACCCCCAAGGAACAAGAGACACTGCTGAGCACCATTAAGTGTGGGCGTCTACAGAGGAATCTGGGGCCTGCTCTGAGCAGGACCTTATCCCAGCTCTACTGCAGCTACGGCCCCTTGACCTGAGCTTATGCCGTGAAACAGCCCCCAGCCCCCGTCTGTCCCCCAACCCAGGCTAGCCAGCCTTTTAACAACATCTTTTGGGGTACAATTAGAATCTAAGTGTTTGCAGCCATATGTGTCATGTAGAAGATGCCTAGCCCTGGGGACCTTGTGAAATACGCAGGAACCAGGGATAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Wai Chan et al.
Nature communications, 5, 4844-4844 (2014-09-12)
Holliday junction (HJ) resolvases are necessary for the processing of persistent recombination intermediates before cell division. Their actions, however, need to be restricted to the late stages of the cell cycle to avoid the inappropriate cleavage of replication intermediates. Control
Yuping Yin et al.
Molecular cancer therapeutics, 18(8), 1439-1450 (2019-05-31)
DNA replication and repair proteins play an important role in cancer initiation and progression by affecting genomic instability. The DNA endonuclease Mus81 is a DNA structure-specific endonuclease, which has been implicated in DNA replication and repair. In this study, we
Delphine Lemaçon et al.
Nature communications, 8(1), 860-860 (2017-10-19)
The breast cancer susceptibility proteins BRCA1 and BRCA2 have emerged as key stabilizing factors for the maintenance of replication fork integrity following replication stress. In their absence, stalled replication forks are extensively degraded by the MRE11 nuclease, leading to chemotherapeutic
Ying Wai Chan et al.
Nature cell biology, 20(1), 92-103 (2017-12-20)
The resolution of joint molecules that link recombining sister chromatids is essential for chromosome segregation. Here, we determine the fate of unresolved recombination intermediates arising in cells lacking two nucleases required for resolution (GEN1 -/- knockout cells depleted of MUS81).
Haiqing Fu et al.
Nature communications, 6, 6746-6746 (2015-04-17)
The Mus81 endonuclease resolves recombination intermediates and mediates cellular responses to exogenous replicative stress. Here, we show that Mus81 also regulates the rate of DNA replication during normal growth by promoting replication fork progression while reducing the frequency of replication

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service