E9383
Antibody Sensitized Sheep Erythrocytes
1 × 109 cells/mL, suitable for assay of complement component activity (H50 units) and whole complement activity (CH50 units), buffered aqueous cell suspension
Synonym(s):
EA7S
About This Item
Recommended Products
form
buffered aqueous cell suspension
drug control
regulated under CDSA - not available from Sigma-Aldrich Canada
concentration
1 × 109 cells/mL
suitability
suitable for assay of complement component activity (H50 units) and whole complement activity (CH50 units)
shipped in
wet ice
storage temp.
2-8°C
General description
Physical form
Other Notes
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service