Skip to Content
Merck
All Photos(1)

Key Documents

C1515

Supelco

Chromosorb® Porous Polymer Adsorbent

matrix (Chromosorb 104), 60-80 mesh, bottle of 50 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
Pricing and availability is not currently available.

packaging

bottle of 50 g

matrix

(Chromosorb 104)

particle size

60-80 mesh

pore size

0.06-0.08 μm average diameter

Looking for similar products? Visit Product Comparison Guide

General description

Chromosorb porous polymers, originally developed and designed for use in packed gas chromaotgraphy (GC) columns, are polyaromatic cross-linked materials with uniform rigid structures. Produced by co-polymerization of monofunctional monomers with difunctional monomers, they are infusible and insoluble in most solvents.

For more information about any of our adsorbents, please visit sigma-aldrich.com/adsorbents

Legal Information

Chromosorb is a registered trademark of Imerys Minerals California, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service