Key Documents
AXYVT3845ULR
Pipette tips in 384-tip format, robotic
size 5 μL, pkg of 5x3840tips/cs, racked
Synonym(s):
pipet tips
Select a Size
1 PKG
HUF 2,120,000.00
HUF 2,120,000.00
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
HUF 2,120,000.00
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
size:
5 μL
packaging:
pkg of 5x3840tips/cs
manufacturer/tradename:
Axygen Inc VT-384-5UL-R
Corning
HUF 2,120,000.00
Please contact Customer Service for Availability
manufacturer/tradename
Axygen Inc VT-384-5UL-R
Corning
packaging
pkg of 5x3840tips/cs
size
5 μL
General description
  • Axygen® automation pipette tips in 384-tip format are designed for Agilent®/Velocity11 VPrep® and Bravo 96LT, 8LT, 384ST, 96ST, and 16ST head.
  • Manufactured to stringent specifications including automated assembly to minimize contamination risks and ensure consistent performance and quality.
  • The clear polypropylene high-quality tips are ideal for sensitive biological applications.
  • RNase-/DNase-free and nonpyrogenic.
Other Notes
Browse all pipette tips
Legal Information
Agilent is a registered trademark of Agilent Technologies, Inc.
Axygen is a registered trademark of Corning, Inc.
Velocity11 VPrep is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)