Skip to Content
Merck
All Photos(2)

Key Documents

AV100832

Sigma-Aldrich

Anti-GABPA antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-GA binding protein transcription factor, α subunit 60 kDa

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
HUF 131,000.00

HUF 131,000.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
HUF 131,000.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

HUF 131,000.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

51 kDa

species reactivity

dog, horse, human, bovine, mouse, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GABPA(2551)

General description

GA-binding protein (GABP) is a member of Ets family of transcription factors that functions as a heterodimer. The α subunit binds to DNA while the β subunit enables transactivation of the target genes.

Immunogen

Synthetic peptide directed towards the C terminal region of human GABPA

Application

Anti-GABPA antibody produced in rabbit is suitable for western blotting at a concentration of 0.5-1 μg/ml. For immunohistochemistry of paraffin-embedded tissue sections, a concentration of 4-8 μg/ml is suitable.

Biochem/physiol Actions

GABPA subunit has a distinct role cell cycle progression, embryogenesis and synaptic function at neuromuscular junctions. It is reported to regulate the cell migration and cytoskeletal changes in breast epithelial cells.

Sequence

Synthetic peptide located within the following region: KWGQRKNKPTMNYEKLSRALRYYYDGDMICKVQGKRFVYKFVCDLKTLIG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zaneta Odrowaz et al.
PloS one, 7(12), e49892-e49892 (2013-01-04)
Members of the ETS transcription factor family often target the same binding regions and hence have the potential to regulate the same genes and downstream biological processes. However, individual family members also preferentially bind to other genomic regions, thus providing
F C de la Brousse et al.
Genes & development, 8(15), 1853-1865 (1994-08-01)
This report outlines three observations relating to GABP beta, a polypeptide constituent of the heterotetrameric transcription factor GABP. Evidence is presented showing that the mouse genome encodes two highly related GABP beta polypeptides, designated GABP beta 1-1 and GABP beta
Xuefang Jing et al.
The Journal of biological chemistry, 283(36), 24326-24333 (2008-07-17)
GA-binding protein (GABP) is the only Ets family transcription factor that functions as a heterodimer. The GABPalpha subunit binds to DNA, and the GABPbeta subunit possesses the ability to transactivate target genes. Inactivation of GABPalpha caused embryonic lethality and defective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service