Skip to Content
Merck
All Photos(1)

Key Documents

51947

Sigma-Aldrich

N-Acetyl-D-galactosamine 6-sulfate sodium salt

≥98.0% (TLC)

Synonym(s):

GalNAc-6S

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H14O9NSNa
CAS Number:
Molecular Weight:
323.25
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

bovine (Ruminant- Cow, Ox, Buffalo)

Quality Level

description

Mixed anomers

Assay

≥98.0% (TLC)

form

solid

optical activity

[α]/D 54.0±4.0, c = 0.1% (w/v) in water

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

−20°C

SMILES string

[Na+].CC(=O)N[C@H]1C(O)O[C@H](COS([O-])(=O)=O)[C@H](O)[C@@H]1O

InChI

1S/C8H15NO9S.Na/c1-3(10)9-5-7(12)6(11)4(18-8(5)13)2-17-19(14,15)16;/h4-8,11-13H,2H2,1H3,(H,9,10)(H,14,15,16);/q;+1/p-1/t4-,5-,6+,7-,8?;/m1./s1

InChI key

CBUJZKTVEFVBBG-DBKUKYHUSA-M

Looking for similar products? Visit Product Comparison Guide

Application

N-Acetyl-D-galactosamine 6-sulfate (GalNAc-6S) is used as a substrate to identify, differentiate and characterize N-acetylgalactosamine sulfatase(s). GalNAc-6S is used to study the rare autosomal recessive disorder Mucopolysaccharidosis IVA (MPS IVA).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Tomatsu et al.
Current pharmaceutical biotechnology, 12(6), 931-945 (2011-04-22)
Mucopolysaccharidosis IVA (MPS IVA), also known as Morquio A, is a rare, autosomal recessive disorder caused by a deficiency of the lysosomal enzyme N-acetylgalatosamine-6-sulfate-sulfatase (GALNS), which catalyzes a step in the catabolism of glycosaminoglycans (GAGs), keratan sulfate (KS) and chondroitin-6-sulfate
Souhir Khedhiri et al.
Diagnostic pathology, 6, 11-11 (2011-01-22)
Mucopolysaccharidosis type IVA or Morquio A syndrome is characterized by the lack of N-acetylgalactosamine-6-sulfate-sulfatase and the accumulation of keratan sulfate and chondroitin-6-sulfate in the lysosomes. At least, 148 mutations and 16 polymorphisms were identified in the GALNS gene.The aim of
G J Gibson et al.
The Biochemical journal, 248(3), 755-764 (1987-12-15)
Initial purification of N-acetylgalactosamine-4-sulphate sulphatase from human liver homogenates containing approx. 1 mg of enzyme in 26 g of soluble proteins was achieved by a six-column chromatography procedure and yielded approx. 40 micrograms of a single major protein species. Enzyme
J Bielicki et al.
The Biochemical journal, 279 ( Pt 2), 515-520 (1991-10-15)
Human N-acetylgalactosamine 6-sulphatase (EC 3.1.6.14), which is involved in the lysosomal degradation of the glycosaminoglycans keratan sulphate and chondroitin 6-sulphate, was purified more than 130,000-fold in 2.8% yield from liver by an eight-step column procedure. One major form was identified
M Masue et al.
Journal of biochemistry, 110(6), 965-970 (1991-12-01)
N-Acetylgalactosamine-6-sulfate sulfatase from human placenta was purified 33,600-fold using beta-N-acetyl-D-galactosamine 6-sulfate-(1----4)-beta-D-glucuronic acid-(1----3)-N-acetyl-D-[3H]galactosaminitol 6-sulfate as the substrate. This enzyme is an oligomer with a molecular mass of 120 kDa and consists of polypeptides of 40 and 15 kDa. The 15 kDa

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service