Skip to Content
Merck
All Photos(1)

Key Documents

39966

Sigma-Aldrich

N-Decanoyl-D-sphingosine

≥98.0% (TLC)

Synonym(s):

(2S,3R,4E)-2-(Decanoylamino)-4-octadecene-1,3-diol, N-Decanoyl-D-erythro-sphingosine, N-[(1S,2R,3E)-2-Hydroxy-1-(hydroxymethyl)-3-heptadecen-1-yl]decanamide, C10 Ceramide (d18:1/10:0), C10-Ceramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H55NO3
CAS Number:
Molecular Weight:
453.74
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.85

Assay

≥98.0% (TLC)

form

powder or solid

composition

carbon content, 74.12%
hydrogen content, 12.22%
nitrogen content, 3.09%

impurities

≤5.0 wt. % water

functional group

amide
hydroxyl

lipid type

sphingolipids

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C28H55NO3/c1-3-5-7-9-11-12-13-14-15-16-18-19-21-23-27(31)26(25-30)29-28(32)24-22-20-17-10-8-6-4-2/h21,23,26-27,30-31H,3-20,22,24-25H2,1-2H3,(H,29,32)/b23-21+/t26-,27+/m0/s1

InChI key

FDWVAQFAMZLCAX-NBNLIBPQSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F G Dupuy et al.
Colloids and surfaces. B, Biointerfaces, 149, 89-96 (2016-10-14)
Ceramides constitute a group of lipids with usually high melting temperature that also favor negative curvature in membranes when mixed with other lipids. The short chain C10:0 ceramide is an asymmetric lipid which consists of an 18 carbon sphingosine base
Meei-Hua Lin et al.
Scientific reports, 9(1), 13254-13254 (2019-09-15)
Fatty acid transport protein 4 (FATP4) is an acyl-CoA synthetase that is required for normal permeability barrier in mammalian skin. FATP4 (SLC27A4) mutations cause ichthyosis prematurity syndrome, a nonlethal disorder. In contrast, Fatp4-/- mice die neonatally from a defective barrier.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service