Skip to Content
Merck
All Photos(1)

Key Documents

938769

Sigma-Aldrich

Potassium carbonate ChemBeads

Synonym(s):

K2CO3

Sign Into View Organizational & Contract Pricing

Select a Size

938769-1G
HUF 31,400.00

HUF 31,400.00


Please contact Customer Service for Availability


Select a Size

Change View
938769-1G
HUF 31,400.00

About This Item

Empirical Formula (Hill Notation):
K2CO3
CAS Number:
Molecular Weight:
138.21
MDL number:
UNSPSC Code:
12352100

HUF 31,400.00


Please contact Customer Service for Availability

form

solid

Quality Level

composition

loading of base, 14-16 wt. %

pH

11-13 (25 °C)

SMILES string

[K+].[K+].[O-]C([O-])=O

InChI

1S/CH2O3.2K/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

BWHMMNNQKKPAPP-UHFFFAOYSA-L

General description

Potassium carbonate is an inorganic base. It participates as a base in the palladacycle catalyzed Heck reaction of chlorobenzene with styrene. Addition of amino acids (glycine, sarcosine and proline) to K2CO3 (solvent) promotes the absorption of CO2.

Application

Potassium carbonate has been used in the preparation of disulfonated bis[4-(3-aminophenoxy)phenyl]sulfone, a disulfonated diamine monomer.

It may be used in the following studies:

  • As a reagent in the synthesis of polysubstituted iodobenzene derivatives.
  • As a base for the synthesis of high molecular weight homopolymers and copolymers derived from various bisphenols.
  • As a base for the Suzuki coupling of aryl halides with aryl boronic acids.
  • The Heck reaction of styrene and bromobenzene.ChemBeads are chemical coated glass beads.
ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

For general uses, product is also available in powdered form (209619)

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service