Skip to Content
Merck
All Photos(1)

Key Documents

90054

Supelco

N-Sulfocarbamoylgonyautoxin-1 and N-Sulfocarbamoylgonyautoxin-4

20 μg/g in acetic acid (nominal concentration), analytical standard

Synonym(s):

N-Sulfocarbamoylgonyautoxin-1 and N-Sulfocarbamoylgonyautoxin-4, C3 and C4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17N7O12S2
Molecular Weight:
491.41
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

20 μg/g in acetic acid (nominal concentration)

format

multi-component solution

storage temp.

−20°C

General description

Paralytic Shellfish Toxin PST for the analysis of marine toxins in seafood.

Check out our complete range of marine toxins reference materials

Analysis Note

20 μg/g in 5 mM acetic acid
the mixture is "not equilibrated".

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service