Skip to Content
Merck
All Photos(1)

Key Documents

82370

Sigma-Aldrich

1-Propanethiol

puriss., ≥98.5% (GC)

Synonym(s):

n-Propylmercaptan, Mercaptan C3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2SH
CAS Number:
Molecular Weight:
76.16
Beilstein:
1696860
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

vapor density

2.54 (vs air)

vapor pressure

122 mmHg ( 20 °C)

grade

puriss.

Assay

≥98.5% (GC)

refractive index

n20/D 1.426 (lit.)
n20/D 1.438

bp

67-68 °C (lit.)

mp

−113 °C (lit.)

density

0.820 g/mL at 25 °C (lit.)

SMILES string

CCCS

InChI

1S/C3H8S/c1-2-3-4/h4H,2-3H2,1H3

InChI key

SUVIGLJNEAMWEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service