Skip to Content
Merck
All Photos(1)

Key Documents

80104

Supelco

Maltodextrin test sticks according to Ph. Eur.

for the detection of glucose in solutions

Synonym(s):

Glucose test sticks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319
NACRES:
NA.21

quality

for the detection of glucose in solutions

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages
general analytical

General description

Maltodextrin is a polysaccharide. It is soluble in water and has a mild sweet taste. It can soak fatty substances, hence is used in food industry.

Application

Test sticks give positive reaction for all reducing sugars
Test sticks may be used to determine blood glucose level using blood from finger tips.[1] It may also be used in analysing urinary glucose.[2]

Packaging

package with 50 sticks

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insulin. Its role in the thermic effect of glucose.
Christin L
The Journal of Clinical Investigation, 77(6), 1747-1755 (1986)
Infrared spectroscopic analysis of human interstitial fluid in vitro and in vivo using FT-IR spectroscopy and pulsed quantum cascade lasers (QCL): Establishing a new approach to non invasive glucose measurement.
Pleitez M
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 85(1), 61-65 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service