Skip to Content
Merck
All Photos(1)

Key Documents

72557

Millipore

EC O157:H7 ChromoSelect Selective Agar, Base

suitable for microbiology

Synonym(s):

EC O157:H7 Selective Agar ChromoSelect, Base

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
bile salts mixture, 1.5 g/L
casein enzymic hydrolysate, 8.0 g/L
chromogenic mixture, 0.25 g/L
sodium lauryl sulfate, 0.1 g/L
sorbitol, 7.0 g/L

final pH

6.8±0.2 (25 °C)

application(s)

agriculture
clinical testing
environmental
food and beverages
food and beverages
food and beverages

microbiology

suitability

selective and differential for Escherichia coli
selective and differential for coliforms
selective and differential for enterovirulent E. coli (EEC)

Application

EC O157:H7 ChromoSelect Selective Agar, Base is recommended for selective isolation and easy detection of Escherichia coli O157:H7 from food samples.

Preparation Note

Supplement:
1 vial/l of ECO157:H7 ChromoSelect Selective Supplement (Cat. No. 44931)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service