Skip to Content
Merck
All Photos(1)

Key Documents

37008

Sigma-Aldrich

Digitonin

~50% (TLC)

Synonym(s):

Digitin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H92O29
CAS Number:
Molecular Weight:
1229.31
Beilstein:
78654
EC Number:
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

description

non-ionic

optical activity

[α]20/D −54.5±3°, c = 1% in methanol (dry matter)

mol wt

micellar avg mol wt 70,000

concentration

~50% (TLC)

aggregation number

60

CMC

<0.5 mM (20-25°C)

mp

230-240 °C (dec.) (lit.)

solubility

ethanol: 10 mg/mL, clear, almost colorless (hot)

SMILES string

C[C@@]12[C@]([C@@H]3C)([H])[C@](O[C@]34CC[C@@H](C)CO4)([H])[C@@H](O)[C@@]1([H])[C@@](CC[C@]5([H])[C@@]6(C[C@@H](O)[C@H](O[C@]([C@@H]([C@@H](O)[C@H]7O[C@@](O[C@H](CO)[C@@H](O)[C@@H]8O[C@@](OC[C@@H](O)[C@@H]9O)([H])[C@@H]9O)([H])[C@@H]8O[C@@](O[C@H](CO)[C@H

InChI

1S/C56H92O29/c1-19-7-10-56(75-17-19)20(2)31-45(85-56)37(67)32-22-6-5-21-11-26(24(61)12-55(21,4)23(22)8-9-54(31,32)3)76-50-42(72)39(69)44(30(16-60)80-50)81-53-48(47(36(66)29(15-59)79-53)83-49-40(70)33(63)25(62)18-74-49)84-52-43(73)46(35(65)28(14-58)78-52)82-51-41(71)38(68)34(64)27(13-57)77-51/h19-53,57-73H,5-18H2,1-4H3/t19-,20+,21+,22-,23+,24-,25-,26-,27-,28-,29-,30-,31+,32-,33+,34-,35+,36-,37+,38+,39-,40-,41-,42-,43-,44+,45-,46+,47+,48-,49+,50-,51+,52+,53+,54-,55+,56-/m1/s1

InChI key

UVYVLBIGDKGWPX-KUAJCENISA-N

Looking for similar products? Visit Product Comparison Guide

Application

suitable for determination of cholesterol

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Permeabilizes plasma membranes of eukaryotic cells by complexing with membrane cholesterol and other unconjugated β-hydroxysterols. Use of digitonin-permeabilized adipocytes for cAMP studies;[1] Selective precipitation of cholesterol;[2] Solubilization of membrane proteins;[3] Opioid receptor proteins.[4]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Scallen et al.
The Journal of cell biology, 40(3), 802-813 (1969-03-01)
A major methodological problem in the intracellular localization of cholesterol is the nearly complete extraction of sterols during routine dehydration and embedding procedures for electron microscopy. Cholesterol digitonide (a sterol complex with digitonin), however, is qualitatively insoluble in these solvents.
C D Demoliou-Mason et al.
FEBS letters, 170(2), 378-382 (1984-05-21)
The binding sites for opiates (agonist and antagonist) and opioid peptides can be solubilized from rat brain membranes with digitonin in the presence of Mg2+ (10 mM). High affinity and high capacity binding to the soluble delta, mu, and kappa
Solubilization of functional membrane proteins.
L M Hjelmeland et al.
Methods in enzymology, 104, 305-318 (1984-01-01)
Use of digitonin-permeabilized adipocytes for cAMP studies.
R A Mooney
Methods in enzymology, 159, 193-202 (1988-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service