Skip to Content
Merck
All Photos(1)

Key Documents

31059

Supelco

α,α-Dichlorotoluene

PESTANAL®, analytical standard

Synonym(s):

(Dichloromethyl)benzene, Benzal chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CHCl2
CAS Number:
Molecular Weight:
161.03
Beilstein:
1099407
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

vapor pressure

0.3 mmHg ( 20 °C)

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.550 (lit.)

bp

82 °C/10 mmHg (lit.)

mp

−17-−15 °C (lit.)

density

1.254 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

ClC(Cl)c1ccccc1

InChI

1S/C7H6Cl2/c8-7(9)6-4-2-1-3-5-6/h1-5,7H

InChI key

CAHQGWAXKLQREW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EG Label

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzal chloride.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 29, 65-72 (1982-05-01)
Occupational exposure to benzyl chloride and benzyl chloride due to contaminated butyl benzyl phthalate.
L Rittfeldt et al.
Scandinavian journal of work, environment & health, 9(4), 367-368 (1983-08-01)
Carlos Eduardo Verdiani de Carvalho et al.
Acta cirurgica brasileira, 21(1), 43-46 (2006-02-24)
To investigate the effects of intrinsic denervation of the jejunum after the extensive intestinal resection in rats. Thirty male Wistar rats were distributed into three groups, depending on the experimental procedure: Group C (control), Group R (resection) and Group D
K Fukuda et al.
Gan, 72(5), 655-664 (1981-10-01)
The carcinogenicity of benzyl chloride (BYC), benzal chloride (BAC), benzotrichloride (BTC) and benzoyl chloride (BOC), which were suspected as causative agents of lung cancer and maxillary malignant lymphoma of workers employed in factories producing BOC, was examined by skin applications
Lung cancer mortality among workers in a factory manufacturing chlorinated toluenes: 1961-84.
T Sorahan et al.
British journal of industrial medicine, 46(6), 425-427 (1989-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service