Skip to Content
Merck
All Photos(1)

Key Documents

16187

Sigma-Aldrich

SODOSIL RA 17

alkaline cleaner free of chlorine

Sign Into View Organizational & Contract Pricing

Select a Size

5 L
HUF 58,700.00

HUF 58,700.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 L
HUF 58,700.00

About This Item

UBA Number:
08050023
UNSPSC Code:
12352000
NACRES:
NA.22

HUF 58,700.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

pH

11.5-13.5 (20 °C, 1%)

density

1.35-1.41 g/cm3 (20°C)

General description

SODOSIL RA 17 is a sterilization and decontamination kit, used to clean lab containers and surfaces for downstream scientific operations involving cell culture and related techniques.

Legal Information

SODOSIL is a trademark of Sigma-Aldrich Chemie GmbH

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service