Skip to Content
Merck
All Photos(1)

Key Documents

10979

Supelco

Polyvinyl alcohol mounting medium with NPG, antifading

pH 8.5-8.9

Synonym(s):

PVA-NPG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
Pricing and availability is not currently available.

refractive index

n20/D 1.378

pH

8.5-8.9

density

1.08 g/mL at 20 °C

suitability

in accordance for application

storage temp.

2-8°C

Physical form

Aqueous solution containing tris(hydroxymethyl)aminomethane, tris(hydroxymethyl)aminomethane hydrochloride, polyvinyl alcohol 22000, glycerol, and propyl gallate

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service