Skip to Content
Merck
All Photos(1)

Key Documents

03938

Sigma-Aldrich

1-Ethyl-3-methylimidazolium bromide

dry, ≥98.5% (HPLC/T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11BrN2
CAS Number:
Molecular Weight:
191.07
Beilstein:
5162084
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39161001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.5% (HPLC/T)

impurities

≤200 ppm water

SMILES string

[Br-].CCn1cc[n+](C)c1

InChI

1S/C6H11N2.BrH/c1-3-8-5-4-7(2)6-8;/h4-6H,3H2,1-2H3;1H/q+1;/p-1

InChI key

GWQYPLXGJIXMMV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service