Skip to Content
Merck
All Photos(1)

Key Documents

28-6080

Sigma-Aldrich

Sulfuric acid, fuming

JIS special grade, 30.0-33.0%

Synonym(s):

Oleum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4 · (SO3)x
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
Pricing and availability is not currently available.

grade

JIS special grade

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 25 °C)

form

liquid

availability

available only in Japan

concentration

30.0-33.0%

density

1.925 g/mL at 25 °C (lit.)

SMILES string

O=S(=O)=O.OS(O)(=O)=O

InChI

1S/H2O4S.O3S/c1-5(2,3)4;1-4(2)3/h(H2,1,2,3,4);

InChI key

HIFJUMGIHIZEPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service