Skip to Content
Merck
All Photos(1)

Key Documents

SX0760E

Supelco

Sodium sulfate

anhydrous, crystals, Meets ACS Specifications Tracepur®

Synonym(s):

Disodium Sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-820-9
grade:
ACS reagent
form:
crystals
solubility:
200 g/L
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

product line

Tracepur®

Assay

≥99.0%

form

crystals

impurities

≤0.01% Insoluble matter
≤1% Mesh size (100 and finer)

loss

≤0.5% loss on ignition

color

white

pH

5.2-8.0 (20 °C, 50 g/L in H2O)

mp

884 °C (lit.)

solubility

200 g/L

density

2.68 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%

cation traces

Ca: ≤0.01%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.005%
N: ≤5 ppm
heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

anhydrous, crystals, Meets ACS Specifications Tracepur®

Analysis Note

Assay99.0% minCalcium (Ca)0.01% maxChloride (Cl)0.001% maxColorWhiteFormCrystalsHeavy Metals (by ICP-OES)5ppm maxInsoluble matter0.01% maxIron (Fe)0.001% maxLoss on ignition0.5% maxMagnesium (Mg)0.005% maxMesh size (10-30)80% minMesh size (10-60)95% minMesh size (100 and finer)1.0% maxNitrogen compounds (as N)5ppm maxPhosphate (PO4)0.001% maxPotassium (K)0.01% maxSuitability for residue analysis (dried 400°/4 hr)To pass testpH of a 5% solution at 25°C5.2-9.2

Legal Information

TRACEPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service