SMWP29325
MF-Millipore Membrane
pore size 5.0 μm, filter diam. 293 mm, hydrophilic
Synonym(s):
MF-Millipore Membrane, mixed cellulose esters, Hydrophilic, 5.0 μm, 293 mm, white, plain
About This Item
Recommended Products
material
mixed cellulose esters (MCE)
plain filter
white filter
Quality Level
feature
hydrophilic
manufacturer/tradename
MF-Millipore™
parameter
32 L/min-cm2 air flow rate
580 mL/min-cm2 water flow rate
75 °C max. temp.
filter diam.
293 mm
thickness
160 μm
gravimetric extractables
6%
refractive index
n/D 1.495
matrix
MF-Millipore™
pore size
5.0 μm pore size
84 % porosity
bubble point
≥0.42 bar, air with water at 23 °C
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH SVHC Candidate List
EU REACH Annex XIV (Authorisation List)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service