MABS1329
Anti-Annexin A3 Antibody, clone 9996-1-3M1/3L7-5
clone 9996-1-3M1/3L7-5, from mouse
Synonym(s):
Annexin A3, 35-alpha calcimedin, Annexin III, Annexin-3, Inositol 1, 2-cyclic phosphate 2-phosphohydrolase, Lipocortin III, PAP-III, Placental anticoagulant protein III
About This Item
biological source
mouse
Quality Level
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
9996-1-3M1/3L7-5, monoclonal
species reactivity
human
technique(s)
immunohistochemistry: suitable (paraffin)
neutralization: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... ANXA3(306)
General description
Specificity
Immunogen
Application
Western Blotting Analysis: 10 µg/mL from a representative lot detected Annexin A3 in 10 µg of human spleen tissue lysate.
Immunohistochemistry Analysis: A representative lot detected a markedly downregulated annexin A3 immunoreactivity in Huh7 xenografts from antibody-treated mice, while an upregulated annexin A3 immunoreactivity was seen in xenografts from Cisplatin-treated mice (Tong, M., et al. (2015). Stem Cell Reports. 5(1):45-59).
Neutralizing Analysis: A representative lot inhibited the proliferation of CD133-/annexin A3-positive Huh7 cells, but not the CD133-/annexin A3-negative MIHA cells. Antibody treatment further sensitized Huh7 to Cisplatin (Cat. No. 232120) antiproliferative activity (Tong, M., et al. (2015). Stem Cell Reports. 5(1):45-59).
Neutralizing Analysis: A representative lot suppressed the migration, invasion, angiogenesis induction, and hepatospheres formation of human hepatocellular carcinoma (HCC) cells (Tong, M., et al. (2015). Stem Cell Reports. 5(1):45-59).
Neutralizing Analysis: A representative lot inhibited the growth of Huh7 xenograph in mice. Residual xenografts following antibody treatment (alone or with Cisplatin), but not Cisplatin treatment alone failed to form tumor mass when serially transplanted into secondary recipients (Tong, M., et al. (2015). Stem Cell Reports. 5(1):45-59).
Western Blotting Analysis: A representative lot detected the expression of the ~33 kDa Annexin A3 among a panel of human hepatocellular carcinoma (HCC) cell lines (Tong, M., et al. (2015). Stem Cell Reports. 5(1):45-59).
Signaling
Quality
Western Blotting Analysis: 5 µg/mL of this antibody detected Annexin A3 in 10 µg of human placenta tissue lysate.
Target description
Physical form
Storage and Stability
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service