Skip to Content
Merck
All Photos(1)

Key Documents

MABF1519

Sigma-Aldrich

Anti-mouse IFN gamma, PE-Cy7, clone XMG1.2 Antibody

clone XMG1.2, from rat, PE-Cy7

Synonym(s):

Interferon gamma (IFN-gamma), IFN gamma, PE-Cy7

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
HUF 51,500.00

HUF 51,500.00


Estimated to ship onApril 11, 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
HUF 51,500.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

HUF 51,500.00


Estimated to ship onApril 11, 2025Details


Request a Bulk Order

biological source

rat

Quality Level

conjugate

PE-Cy7

antibody form

purified antibody

antibody product type

primary antibodies

clone

XMG1.2, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ifng(15978)

General description

The XMG1.2 antibody is specific for mouse Interferon-gamma (IFN-g), a 20 kDa type II cytokine known for its central roles in protection against bacterial or viral pathogens and for its anti-tumor properties. IFN-g is secreted by several types of immune cells which allow the cytokine to modulate innate immunity when secreted by NK and NKT cells, and to function in support of adaptive immunity when secreted by Th1 and CD8+ T cells (CTLs).The XMG1.2 antibody is suitable for detection of intracellular IFN-g protein by flow cytometry. Other formats can be used for quantitative analysis of the secreted protein by ELISA when paired with an appropriate capture antibody. This clone has been reported for neutralization of the functional activity of IFN-g in a variety of assays (use format suitable for functional assays).

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Anti-mouse IFN gamma, PE-Cy7, clone XMG1.2 Antibody is an antibody against mouse IFN gamma for use in Flow Cytometry.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.125 μg from a representative lot detected IFN gamma, PE-Cy7 in one million C57Bl/6 splenocytes.

Physical form

Purified Rat monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2.
Purified by size-exclusion chromatography

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service