Skip to Content
Merck
All Photos(3)

Key Documents

MABE150

Sigma-Aldrich

Anti-KDM5B Antibody, clone 15G8.1

clone 15G8.1, from mouse

Synonym(s):

Lysine-specific demethylase 5B, KDM5B, Cancer/testis antigen 31, CT31, Histone demethylase JARID1B, Jumonji/ARID domain-containing protein 1B, PLU-1, Retinoblastoma-binding protein 2 homolog 1, RBP2-H1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
HUF 143,000.00

HUF 143,000.00


Estimated to ship onApril 15, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
HUF 143,000.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

HUF 143,000.00


Estimated to ship onApril 15, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

15G8.1, monoclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KDM5B(10765)

General description

Lysine-specific demethylase 5B (UniProt Q9UGL1; also known as Cancer/testis antigen 31, CT31, Histone demethylase JARID1B, Jumonji AT rich interactive domain 1B, Jumonji/ARID domain-containing protein 1B, PLU-1, Protein phosphatase 1 regulatory subunit 98, Putative DNA/chromatin binding motif, RBP2-H1, Retinoblastoma-binding protein 2 homolog 1) is encoded by the KDM5B (also known as PLU1, PUT1, JARID1B, PPP1R98, RBBP2H1, RBBP2H1A) gene (Gene ID 10765) in human. JARID1B/KDM5B is an H3K4me2/3 demethylase that regulates self-renewal and differentiation in a cell type-specific manner. JARID1B/KDM5B is highly expressed in embryonic stem cells (ESCs), hematopoietic stem cells (HSCs), and in various primary cancers including melanoma, breast, testicular, and is upregulated in leukemic cell lines.

Immunogen

Epitope: Region between ARID and JmjC domain
GST-tagged recombinant protein corresponding to the region between ARID and JmjC domain of human KDM5B.

Application

This Anti-KDM5B Antibody, clone 15G8.1 is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of KDM5B.
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected KDM5B in 10 µg of MCF-7 cell lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected KDM5B in human kidney and mouse cerebral cortex tissue.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected KDM5B in 10 µg of HeLa cell lysate.

Target description

~175 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service