Skip to Content
Merck
All Photos(2)

Key Documents

MABC1609

Anti-AIF Antibody, clone 3C11

Synonym(s):

Apoptosis-inducing factor 1, mitochondrial, Programmed cell death protein 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

ambient

Gene Information

human ... AIFM1(9131)

General description

Apoptosis-inducing factor 1, mitochondrial (UniProt: O95831; also known as Programmed cell death protein 8) is encoded by the AIFM1 (also known as AIF, PDCD8) gene (Gene ID: 9131) in human. AIF is a nuclear encoded flavoprotein that is generally confined to the mitochondrial intermembrane space and functions both as NADH oxidoreductase and as regulator of apoptosis. Six isoformsi of AIF have been described that are produced by alternative splicing. Under normal conditions, a 54-residue N-terminal (transit peptide) segment is first proteolytically removed during or just after translocation into the mitochondrial intermembrane space (IMS) by the mitochondrial processing peptidase to form the inner-membrane-anchored mature form (AIFmit). Upon induction of apoptosis, it is further proteolytically processed at to generate the mature form, which is confined to the mitochondrial IMS in a soluble form (AIFsol). AIFsol is released to the cytoplasm in response to specific death signals, and translocated to the nucleus, where it induces nuclear apoptosis in a caspase-independent manner. AIF also functions as an anti-apoptotic factor in normal mitochondria via its NADH oxidoreductase activity. Once released from mitochondria it translocates to the nucleus to induce caspase-independent fragmentation of chromosomal DNA. Mutations in AIF gene are linked to combined oxidative phosphorylation deficiency 6 that is characterized by psychomotor delay, hypotonia, areflexia, and muscle weakness and wasting. Defects in this gene are also known to cause Cowchock syndrome that leads to early childhood onset of a slowly progressive axonal sensorimotor neuropathy.

Specificity

Clone 3C11 specificallly detects AIF in human cells.

Immunogen

His-tagged full length recombinant human AIF.

Application

Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected AIF in human skin, human testis, and human kidney tissue.
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in A431 cell lysates.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected AIF in 10 µg of A431 cell lysates.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service