Skip to Content
Merck
All Photos(1)

Key Documents

CB1047

Sigma-Aldrich

Anti-STMN1 Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Stathmin 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
HUF 136,000.00

HUF 136,000.00


Estimated to ship onMarch 18, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
HUF 136,000.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

HUF 136,000.00


Estimated to ship onMarch 18, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

mouse, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Stmn1(16765)

General description

Purified rabbit polyclonal antibody. Recognizes the ~20 kDa STMN1 protein.
Recognizes the ~20 kDa STMN1 protein in mouse spleen tissue.
This Anti-STMN1 Rabbit pAb is validated for use in Immunoblotting for the detection of STMN1.

Immunogen

Full-length, human STMN1 protein (aa 1-149)
Human

Application


Immunoblotting (1-5 g/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Negative Control
293T cells
Positive Control
Mouse spleen tissue

Other Notes

Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chia-Yen Hsiao et al.
Evidence-based complementary and alternative medicine : eCAM, 2012, 467531-467531 (2012-04-27)
Angelica sinensis (AS) is a traditional Chinese herbal medicine that has been formulated clinically to treat various form of skin trauma and to help wound healing. However, the mechanism by which it works remains a mystery. In this study we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service