Skip to Content
Merck
All Photos(1)

Key Documents

AB5401

Sigma-Aldrich

Anti-Growth Associated Protein 43 Antibody, phosphoSer 41

Chemicon®, from rabbit

Synonym(s):

GAP-43, Neuromodulin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
HUF 145,000.00

HUF 145,000.00


Estimated to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
HUF 145,000.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

HUF 145,000.00


Estimated to ship onApril 07, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GAP43(2596)

General description

GAP-43, a common marker of differentiating neurons, is expressed at elevated levels by developing or regenerating neurons during axon growth. While GAP-43 is an integral membrane protein associated with the cytoplasmic surface of axonal growth cones (and synapses), it is absent from dendritic growth cones. In adult brain, GAP-43 is found in high concentration in presynaptic areas where memory formation is thought to occur, such as frontal cortex, limbic system and hippocampus. Phosphorylation of GAP-43 by PKC (at Ser41 on human, mouse, rat, and bovine GAP-43 or Ser42 on chicken and Xenopus GAP-43) is specifically correlated with certain forms of synaptic plasticity.

Specificity

GAP-43 phosphoSer41. The labeling of the GAP-43 protein (~43kDa) in Western blots of rat brain is blocked by the phosphopeptide but not by the dephosphopeptide. In some tissues the antibody may recognize a lower molecular weight band. This band is also detected by a pan GAP-43 antibody.

Immunogen

Epitope: phosphoSer 41
Synthetic peptide corresponding to amino acids surrounding the phosphoSer41 of rat GAP-43. This is the site phosphorylated by protein kinase C.

Application

Anti-Growth Associated Protein 43 Antibody, phosphoSer 41 is an antibody against Growth Associated Protein 43 for use in WB.
Research Category
Neuroscience
Research Sub Category
Neuroregenerative Medicine

Signaling Neuroscience
Western blot: 1:1,000 (43kDa band).

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin in 10 mM HEPES (pH 7.5), 150 mM NaCl with 100 μg/mL BSA and 50% glycerol.

Storage and Stability

Maintain at -20°C in undiluted for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles. Do not store in a self defrosting freezer.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service