Skip to Content
Merck
All Photos(1)

Key Documents

AB15794

Sigma-Aldrich

Anti-Dexras1 Antibody

Chemicon®, from rabbit

Synonym(s):

Activator of G-Protein Signaling 1, AGS1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
HUF 183,000.00

HUF 183,000.00


Estimated to ship onMarch 04, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
HUF 183,000.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

HUF 183,000.00


Estimated to ship onMarch 04, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

rat (100% immunogen homology), human (100% immunogen homology)

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... RASD1(51655)

General description

Dexras1 is a member of the superfamily of RAS-g-proteins and directly binds to Gialpha heterotrimeric G-proteins where it appears to promote guanine nucleotide exchange on Gialpha. Dexras1 is implicated in NMDA receptor signaling through nitric oxide synthase.

Specificity

Dexras1 (Activator of G-Protein Signaling 1, AGS1). By Western blot the antibody recognizes a band at ~33 kDa on mouse brain extract. The antibody has also been used to detect Dexras1 in brown adipocyte lysate.
Other species have not been tested

Immunogen

Synthetic peptide.

Application

Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience
This Anti-Dexras1 Antibody is validated for use in WB for the detection of Dexras1.
Western blot: 1:1,000 on mouse brain extract.
Optimal working dilutions must be determined by the end user.

Physical form

ImmunoAffinity Purified
Liquid in PBS. Contains no preservative

Storage and Stability

6 months at -70°C from date of shipment

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marek Lenarczyk et al.
Journal of molecular histology, 46(2), 123-136 (2014-12-24)
Activators of G-protein Signaling (AGS) are a family of accessory proteins that were discovered as modulators of heterotrimeric G-protein subunits. The primary aim of the present study was to localize Group I and II AGS proteins and determine the renal
Sudhir G Tattikota et al.
Molecular & cellular proteomics : MCP, 12(5), 1214-1225 (2013-01-30)
Argonaute2 (Ago2) is an established component of the microRNA-induced silencing complex. Similar to miR-375 loss-of-function studies, inhibition of Ago2 in the pancreatic β-cell resulted in enhanced insulin release underlining the relationship between these two genes. Moreover, as the most abundant
Li-Juan Zhu et al.
Journal of neurochemistry, 146(5), 598-612 (2018-06-03)
Anxiety disorders are associated with a high social burden worldwide. Recently, increasing evidence suggests that nuclear factor kappa B (NF-κB) has significant implications for psychiatric diseases, including anxiety and depressive disorders. However, the molecular mechanisms underlying the role of NF-κB
Yu Zhang et al.
Aging cell, 17(3), e12754-e12754 (2018-03-27)
In neurons, increased protein-protein interactions between neuronal nitric oxide synthase (nNOS) and its carboxy-terminal PDZ ligand (CAPON) contribute to excitotoxicity and abnormal dendritic spine development, both of which are involved in the development of Alzheimer's disease. In models of Alzheimer's
Sudhir G Tattikota et al.
Cell metabolism, 19(1), 122-134 (2013-12-24)
Pancreatic β cells adapt to compensate for increased metabolic demand during insulin resistance. Although the microRNA pathway has an essential role in β cell proliferation, the extent of its contribution is unclear. Here, we report that miR-184 is silenced in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service