Skip to Content
Merck
All Photos(2)

Key Documents

AB15610-I

Sigma-Aldrich

Anti-Prohormone Convertase PC2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

KEX2-like endoprotease 2, NEC 2, PC2, Prohormone convertase 2, Proprotein convertase 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
HUF 119,000.00

HUF 119,000.00


Estimated to ship onMarch 07, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
HUF 119,000.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

HUF 119,000.00


Estimated to ship onMarch 07, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PCSK2(5126)

General description

Neuroendocrine convertase 2 (EC 3.4.21.94; UniProt P16519; also known as KEX2-like endoprotease 2, NEC 2, PC2, Prohormone convertase 2, Proprotein convertase 2) is encoded by the PCSK2 (also known as NEC2) gene (Gene ID 5126) in human. Neuroendocrine convertase 2/PC2 belongs to the family of subtilisin-related proteolytic enzymes involved in the proteolytic cleavages of peptide hormone precursors. For example, during the multistep processing of the peptide precursor proopiomelanocortin (POMC), PC2 catalyzes the cleavage of adrenocorticotropic hormone 1–39 (ACTH) into adrenocorticotropic hormone 1–14, which is then further processed to produce α-melanocyte-stimulating hormone (α-MSH). PC2 is produced initially in a prepro-form with a signal peptide (a.a. 1-25) and a propeptide (a.a. 26-109) sequence, the removal of which yields the mature 529-amino acid enzyme (a.a. 110-638). The neuroendocrine protein 7B2 blocks proPC2 from forming unactivatable aggregates and is absolutely required for processing proCP2 into active enzyme. Newly synthesized proPC2 is N-glycosylated in the ER and undergoes rate-limiting folding process before it can bind 7B2 and be transported to the TGN/ secretory granules for maturation by sulfation and proteolytic cleavage.

Specificity

This polyclonal antibody detects the ~68 kDa secreted mature PC2 in mouse cerebellum lysate by Western blotting. Expected to recognize also the Prepro- and pre-forms of all three spliced human isoforms reported by UniProt (P16519).

Immunogen

Epitope: Near the N-terminus of mature PC2.
GST-tagged recombinant human mature PC2 N-terminal fragment.

Application

Anti-Prohormone Convertase PC2 Antibody is an antibody against Prohormone Convertase PC2 for use in Western Blotting, Immunohistochemistry.
Immunohistochemistry Analysis: An 1:1,000 dilution from a representative lot detected Prohormone Convertase PC2 in human cerebral cortex, pancreas and testis tissue sections.
Research Category
Neuroscience
Research Sub Category
Hormones & Receptors

Quality

Evaluated by Western Blotting in mouse cerebellum tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Prohormone Convertase PC2 in 10 µg of mouse cerebellum tissue lysate.

Target description

~68 kDa calculated. Target band appears larger than the calculated molecular weight of the mature form (58.28 kDa/mouse & 58.00 kDa/human) due to posttranslational glycosylation and sulfation. 67.97/63.72/68.12 kDa (human isoform 1/2/3 proforms), 70.57/66.31/68.58 kDa (human isoform 1/2/3 prepro-forms) calculated.

Linkage

Replaces: AB15610

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service