Skip to Content
Merck
All Photos(1)

Key Documents

8.45140

Sigma-Aldrich

3,4-Dimethoxybenzoic acid

Msynthplus

Synonym(s):

3,4-Dimethoxybenzoic acid, Veratric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3,4-(CH3O)2C6H3COOH
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352106
EC Index Number:
202-215-7

Assay

≥98.0% (GC)

form

powder

mp

179-182 °C

solubility

0.5 g/L

storage temp.

2-30°C

InChI

1S/C9H10O4/c1-12-7-4-3-6(9(10)11)5-8(7)13-2/h3-5H,1-2H3,(H,10,11)

InChI key

DAUAQNGYDSHRET-UHFFFAOYSA-N

Analysis Note

Colour (visual): white
Appearance of substance (visual): powder
Assay (GC, area%): ≥ 98.0 % (a/a)
Water (K. F.): ≤ 1.0 %
Identity (IR): passes test
1,2-dichloroethane: lot specific result

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service