Skip to Content
Merck
All Photos(1)

Key Documents

8.24420

Sigma-Aldrich

Acetylacetonato(1,5-cyclooctadiene)rhodium(I)

for synthesis

Synonym(s):

Acetylacetonato(1,5-cyclooctadiene)rhodium(I)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19O2Rh
CAS Number:
Molecular Weight:
310.19
MDL number:
UNSPSC Code:
12352103

Quality Level

form

granular

mp

138-140 °C

storage temp.

2-30°C

InChI

1S/C8H12.C5H8O2.Rh/c1-2-4-6-8-7-5-3-1;1-4(6)3-5(2)7;/h1-2,7-8H,3-6H2;3,6H,1-2H3;/b2-1-,8-7-;4-3-;

InChI key

BUYVJWVYKPKZEX-DWVXZKBMSA-N

Analysis Note

Assay (to C, H): ≥ 96.0 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service