Skip to Content
Merck
All Photos(1)

Key Documents

76020USBC

Millipore

GDS RG Power Cord US

BioControl, for use with GDS Rotor-Gene®

Synonym(s):

Power cable, Thermocycler Charger Cord

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
HUF 3,440.00

HUF 3,440.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EACH
HUF 3,440.00

About This Item

UNSPSC Code:
41122409

HUF 3,440.00


Please contact Customer Service for Availability

Request more information

AC/DC input

100 - 200 V AC, 50 - 60 Hz

manufacturer/tradename

BioControl

technique(s)

PCR: suitable

application(s)

food and beverages

compatibility

for use with GDS Rotor-Gene®

storage temp.

room temp

General description

GDS RG Power Cord is the accessory of the Assurance® GDS system. Assurance® GDS is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples. The power cord United States standard connector with 100 - 240 V, 50-60 Hz enables efficient functioning of the GDS system.

Application

GDS RG Power Cord is used to power the GDS Rotor-Gene® Q Thermocycler which is used for the detection of pathogenic organisms in foods, ingredients, and environmental samples.

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Rotor-Gene is a registered trademark of Qiagen GmbH

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service