Key Documents
69739
Sigma-Aldrich
pET-20b(+) DNA - Novagen
Novagen′s pET-20b(+) vector carries an N-terminal pelB signal sequence for potential periplasmic localization
Synonym(s):
cloning vector, expression vector, vector
Select a Size
10 μG
HUF 92,800.00
HUF 92,800.00
Estimated to ship onMarch 04, 2025Details
Request a Bulk Order
Select a Size
Change View
10 μG
HUF 92,800.00
About This Item
UNSPSC Code:
41106610
NACRES:
NA.51
HUF 92,800.00
Estimated to ship onMarch 04, 2025Details