Skip to Content
Merck
All Photos(1)

Key Documents

6.18549

Sigma-Aldrich

n-Heptane

for chromatography

Synonym(s):

Heptane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH3
CAS Number:
Molecular Weight:
100.20
Beilstein:
1730763
MDL number:
UNSPSC Code:
12352001
Pricing and availability is not currently available.

grade

for chromatography

Quality Level

vapor density

3.5 (vs air)

vapor pressure

40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)

autoignition temp.

433 °F

expl. lim.

7 %

refractive index

n20/D 1.387 (lit.)

bp

98 °C (lit.)

mp

−91 °C (lit.)

density

0.684 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC

InChI

1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3

InChI key

IMNFDUFMRHMDMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

n-Heptane is a hydrocarbon liquid commonly usedas a nonpolar mobile phase or solvent in gas chromatography (GC) and reversed-phase liquid chromatography (RPLC).[1][2]

Application


  • Identification of Different Classes of VOCs Based on Optical Emission Spectra Using a Dielectric Barrier Helium Plasma Coupled with a Mini Spectrometer.: This paper describes a novel analytical technique for identifying volatile organic compounds, including n-heptane, demonstrating the versatility of plasma spectroscopy in chemical analysis (Mao et al., 2024).

  • Rapid online analysis of n-alkanes in gaseous streams via APCI mass spectrometry.: This research discusses a rapid analytical method for detecting n-alkanes like n-heptane in gaseous streams, showcasing advancements in mass spectrometry for real-time environmental monitoring (Wentrup et al., 2024).


Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatograph applications in petroleum hydrocarbon fluids
Zeng H, et al.
Advanced gas chromatography-Progress in agricultural, biomedical and industrial applications, 363-388 (2012)
Investigation of the effects of solvent-mismatch and immiscibility in normal-phase ? aqueous reversed-phase liquid chromatography
Groeneveld G, et al.
Journal of Chromatography A, 462818, 1665-1665 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service