Skip to Content
Merck
All Photos(1)

Key Documents

328005

Sigma-Aldrich

ERK Activation Inhibitor Peptide II, Cell-Permeable

The ERK Activation Inhibitor Peptide II, Cell-Permeable controls the biological activity of ERK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

ERK Activation Inhibitor Peptide II, Cell-Permeable, MTP TAT-G-MEK1₁₃, H- GYGRKKRRQRRR-G-MPKKKPTPIQLNP-NH₂, Erk Inhibitor V

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
HUF 109,700.00

HUF 109,700.00


Estimated to ship onMay 05, 2025Details



Select a Size

Change View
1 MG
HUF 109,700.00

About This Item

Empirical Formula (Hill Notation):
C136H241N53O31S
Molecular Weight:
3146.77
UNSPSC Code:
12352200
NACRES:
NA.77

HUF 109,700.00


Estimated to ship onMay 05, 2025Details


Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

water: 1 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable and reversible 13-amino acid peptide corresponding to the N-terminus of MAPK/ERK kinase (MEK or MAPKK) that is fused to the HIV-TAT membrane translocation peptide (MTP) sequence via a glycine linker. Acts as a specific inhibitor of ERK activation by MEK1 and the downstream transcriptional activity of ELK1. Selectively binds to ERK2 and prevents its interaction with MEK (IC50 = 210 nM in vitro). Potently blocks ERK in PMA-stimulated NIH3T3 cells and NGF-treated PC12 cells (IC50 = 29 µM). Does not inhibit the activation of c-jun N-terminal kinases (JNKs) or p38 MAP kinase.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MEK
Product does not compete with ATP.
Reversible: yes
Target IC50: 210 nM against MEK; 29 µM in blocking ERK activation in PMA-stimulated NIH 3T3 cells and NGF-treated PC12 cells

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Gly-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-Gly-Met-Pro-Lys-Lys-Lys-Pro-Thr-Pro-Ile-Gln-Leu-Asn-Pro-NH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kelemen, B.R., et al. 2002. J. Biol. Chem.277, 8741.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martha M Monick et al.
Journal of immunology (Baltimore, Md. : 1950), 177(3), 1636-1645 (2006-07-20)
Human alveolar macrophages, central to immune responses in the lung, are unique in that they have an extended life span in contrast to precursor monocytes. We have shown previously that the ERK MAPK (ERK) pathway is constitutively active in human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service